siRNA Details
| Gene Symbol: LOC105369921 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135015.1 | 2 | 243 | |||
| GenBank mRNA | BX107766.1 | - | 2 | 202 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000551893.1 | - | 2 | 202 | Chr. 12 - 95858237 - 95858798 [-] on Build GRCh38 | |

LOC105369921
Genomic Map