siRNA Details
| Gene Symbol: LOC101928123 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135248.1 | 2 | 664 | |||
| GenBank mRNA | BC044305.1 | - | 2 | 642 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000554614.2 | - | 2 | 642 | Chr. 14 - 73244018 - 73245979 [-] on Build GRCh38 | |

LOC101928123
Genomic Map