siRNA Details
| Gene Symbol: MGAT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BQ023756.1 | - | 1 | 137 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555043.1 | - | 1 | 119 | Chr. 14 - 49620815 - 49623480 [-] on Build GRCh38 | |

MGAT2
Genomic Map