siRNA Details
Gene Symbol: LOC100506999 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | BC038791.1 | - | 2 | 263 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553785.1 | - | 2 | 263 | Chr. 14 - 95573605 - 95578684 [+] on Build GRCh38 |
