siRNA Details
| Gene Symbol: LOC100506999 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC038791.1 | - | 2 | 1026 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000553785.1 | - | 2 | 1026 | Chr. 14 - 95573605 - 95578684 [+] on Build GRCh38 | |

LOC100506999
Genomic Map