siRNA Details
| Gene Symbol: LOC101929718 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110139.1 | 3 | 677 | |||
| GenBank mRNA | AK026285.1 | - | 3 | 677 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000557335.1 | - | 3 | 677 | Chr. 14 - 20995837 - 20999163 [-] on Build GRCh38 | |

LOC101929718
Genomic Map