siRNA Details
| Gene Symbol: LOC105370652 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | JX073282.1 | - | 2 | 404 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000557072.1 | - | 2 | 317 | Chr. 14 - 98136074 - 98139242 [+] on Build GRCh38 | |

LOC105370652
Genomic Map