siRNA Details
| Gene Symbol: CRAT37 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110106.1 | 4 | 867 | |||
| GenBank mRNA | BC036442.1 | - | 4 | 874 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000554333.1 | - | 4 | 869 | Chr. 15 - 91463337 - 91494850 [+] on Build GRCh38 | |

CRAT37
Genomic Map