siRNA Details
Gene Symbol: CRAT37 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110106.1 | 4 | 1083 | |||
GenBank mRNA | BC036442.1 | - | 4 | 1090 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000554333.1 | - | 4 | 1085 | Chr. 15 - 91463337 - 91494850 [+] on Build GRCh38 |
