siRNA Details
| Gene Symbol: LOC254028 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_122043.1 | 3 | 427 | |||
| GenBank mRNA | AK093210.1 | - | 2 | 372 | ||
| BI830942.1 | - | 3 | 432 | |||
| DB034207.1 | - | 3 | 376 | |||
| DB053670.1 | - | 3 | 498 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000554993.1 | - | 2 | 372 | Chr. 14 - 20590402 - 20607137 [+] on Build GRCh38 | |

LOC254028
Genomic Map