siRNA Details
| Gene Symbol: LOC100506498 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135238.1 | 2 | 879 | |||
| GenBank mRNA | BC013914.1 | - | 2 | 824 | ||
| BC013921.1 | - | 2 | 824 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555539.1 | - | 2 | 879 | Chr. 14 - 73822559 - 73830135 [-] on Build GRCh38 | |

LOC100506498
Genomic Map