siRNA Details
| Gene Symbol: LOC101929080 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110127.1 | 3 | 315 | |||
| GenBank mRNA | AK058134.1 | - | 3 | 315 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000554522.1 | - | 3 | 315 | Chr. 14 - 95329584 - 95335504 [+] on Build GRCh38 | |

LOC101929080
Genomic Map