siRNA Details
Gene Symbol: LINC00930 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_021493.1 | 3 | 1243 | |||
GenBank mRNA | AK093148.1 | - | 1088 | |||
AL045331.1 | - | 3 | 443 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000557075.5 | - | 3 | 1022 | Chr. 15 - 92567818 - 92572042 [-] on Build GRCh38 |
