siRNA Details
| Gene Symbol: LINC00930 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_021493.1 | 4 | 1308 | |||
| GenBank mRNA | AK093148.1 | - | 1153 | |||
| AL045331.1 | - | 4 | 508 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000557075.5 | - | 4 | 1087 | Chr. 15 - 92567818 - 92572042 [-] on Build GRCh38 | |

LINC00930
Genomic Map