siRNA Details
| Gene Symbol: LOC100506700 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109999.1 | 3 | 580 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555001.1 | - | 3 | 580 | Chr. 14 - 81441987 - 81450157 [-] on Build GRCh38 | |

LOC100506700
Genomic Map