siRNA Details
| Gene Symbol: LINC00871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102700.1 | 1, 2 | 157 | |||
| GenBank mRNA | BG188315.1 | - | 1, 2 | 158 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC00871| Gene Symbol: LINC00871 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102700.1 | 1, 2 | 157 | |||
| GenBank mRNA | BG188315.1 | - | 1, 2 | 158 | ||