siRNA Details
Gene Symbol: LINC01193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_040094.1 | 5 | 2192 | |||
GenBank mRNA | BC047459.1 | - | 5 | 2192 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000553416.5 | - | 5 | 2192 | Chr. 15 - 20940438 - 20993303 [+] on Build GRCh38 |
