siRNA Details
| Gene Symbol: LOC102724814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110555.1 | 1 | 658 | |||
| GenBank mRNA | AK092855.1 | - | 1 | 658 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555968.1 | - | 1 | 658 | Chr. 14 - 23561097 - 23568073 [+] on Build GRCh38 | |

LOC102724814
Genomic Map