siRNA Details
| Gene Symbol: LOC102724814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110555.1 | 3 | 2255 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC102724814| Gene Symbol: LOC102724814 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110555.1 | 3 | 2255 | |||