siRNA Details
| Gene Symbol: LINC00637 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038436.1 | 3 | 1293 | |||
| GenBank mRNA | AK057986.1 | - | 3 | 1295 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000556586.1 | - | 3 | 1293 | Chr. 14 - 103847721 - 103858049 [+] on Build GRCh38 | |

LINC00637
Genomic Map