siRNA Details
Gene Symbol: LOC105370473 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135257.1 | 3 | 318 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000555433.1 | - | 3 | 318 | Chr. 14 - 44507409 - 44538513 [+] on Build GRCh38 |
