siRNA Details
Gene Symbol: LINC01580 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_120322.1 | 3 | 269 | |||
GenBank mRNA | BC021741.1 | - | 3 | 274 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000555364.5 | - | 3 | 269 | Chr. 15 - 93900701 - 93973822 [+] on Build GRCh38 |
