siRNA Details
| Gene Symbol: LINC01580 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120322.1 | 4 | 455 | |||
| GenBank mRNA | BC021741.1 | - | 4 | 460 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555364.5 | - | 4 | 455 | Chr. 15 - 93900701 - 93973822 [+] on Build GRCh38 | |

LINC01580
Genomic Map