siRNA Details
| Gene Symbol: ZBTB25 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006720250.3 | XP_006720313.1 | 4 | 3109 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000556640.1 | - | 1 | 173 | Chr. 14 - 64422935 - 64448557 [-] on Build GRCh38 | |

ZBTB25
Genomic Map