siRNA Details
| Gene Symbol: LINC01146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046094.1 | 3, 4 | 311 | |||
| GenBank mRNA | BC029479.1 | - | 3, 4 | 311 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01146| Gene Symbol: LINC01146 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_046094.1 | 3, 4 | 311 | |||
| GenBank mRNA | BC029479.1 | - | 3, 4 | 311 | ||