siRNA Details
| Gene Symbol: LINC01599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC084561.1 | - | 1 | 280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01599| Gene Symbol: LINC01599 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BC084561.1 | - | 1 | 280 | ||