siRNA Details
| Gene Symbol: CTAGE5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BM929588.1 | - | 1 | 333 | ||
| Gene Symbol: LOC100288846 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_038935.1 | 1 | 64 | |||
| GenBank mRNA | BC012360.1 | - | 1 | 64 | ||

CTAGE5
Genomic Map