siRNA Details
| Gene Symbol: LOC101927418 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110050.1 | 2, 3 | 252 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555157.1 | - | 2, 3 | 198 | Chr. 14 - 44898908 - 44911863 [-] on Build GRCh38 | |

LOC101927418
Genomic Map