siRNA Details
| Gene Symbol: LINC00519 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | BG121773.1 | - | 1 | 121 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000557518.1 | - | 1 | 121 | Chr. 14 - 51304416 - 51328386 [-] on Build GRCh38 | |

LINC00519
Genomic Map