siRNA Details
| Gene Symbol: LINC01467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110074.1 | 3 | 1005 | |||
| GenBank mRNA | AK130854.1 | - | 4 | 589 | ||
| BC029835.1 | - | 3 | 1005 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LINC01467| Gene Symbol: LINC01467 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_110074.1 | 3 | 1005 | |||
| GenBank mRNA | AK130854.1 | - | 4 | 589 | ||
| BC029835.1 | - | 3 | 1005 | |||