siRNA Details
Gene Symbol: LINC01467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110074.1 | 2 | 741 | |||
GenBank mRNA | BC029835.1 | - | 2 | 741 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000554211.1 | - | 2 | 741 | Chr. 14 - 81605347 - 81623061 [-] on Build GRCh38 |
