siRNA Details
Gene Symbol: LINC01467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110074.1 | 2 | 939 | |||
GenBank mRNA | AK130854.1 | - | 3 | 523 | ||
BC029835.1 | - | 2 | 939 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LINC01467 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_110074.1 | 2 | 939 | |||
GenBank mRNA | AK130854.1 | - | 3 | 523 | ||
BC029835.1 | - | 2 | 939 |