siRNA Details
Gene Symbol: LOC101927351 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK123685.1 | - | 4 | 2386 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000556405.1 | - | 4 | 2385 | Chr. 14 - 44763157 - 44782829 [-] on Build GRCh38 |
