siRNA Details
| Gene Symbol: LOC100506476 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_109995.1 | 3 | 328 | |||
| GenBank mRNA | BM509577.1 | - | 3 | 318 | ||
| CK822534.1 | - | 3 | 328 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000556194.1 | - | 3 | 328 | Chr. 14 - 73787360 - 73803217 [+] on Build GRCh38 | |

LOC100506476
Genomic Map