siRNA Details
| Gene Symbol: LOC100507277 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135240.1 | 1, 2 | 188 | |||
| GenBank mRNA | BU852213.1 | - | 1, 2 | 191 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555725.1 | - | 1, 2 | 188 | Chr. 14 - 101442076 - 101444315 [+] on Build GRCh38 | |

LOC100507277
Genomic Map