siRNA Details
| Gene Symbol: LINC00911 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_102737.1 | 3 | 204 | |||
| NR_102738.1 | 2 | 94 | ||||
| GenBank mRNA | AK131026.1 | - | 3 | 204 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555272.1 | - | 3 | 204 | Chr. 14 - 85393879 - 85420074 [+] on Build GRCh38 | |

LINC00911
Genomic Map