siRNA Details
| Gene Symbol: LINC00648 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_039996.1 | 3 | 924 | |||
| GenBank mRNA | AK056212.1 | - | 3 | 924 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555985.5 | - | 3 | 924 | Chr. 14 - 47764954 - 47795014 [-] on Build GRCh38 | |

LINC00648
Genomic Map