siRNA Details
| Gene Symbol: LINC00644 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_104063.1 | 2 | 259 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000555156.1 | - | 2 | 259 | Chr. 14 - 62134149 - 62139973 [-] on Build GRCh38 | |

LINC00644
Genomic Map