siRNA Details
| Gene Symbol: LOC104613533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126501.1 | 1 | 280 | |||
| GenBank mRNA | BX089571.1 | - | 1 | 280 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

LOC104613533| Gene Symbol: LOC104613533 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_126501.1 | 1 | 280 | |||
| GenBank mRNA | BX089571.1 | - | 1 | 280 | ||