siRNA Details
| Gene Symbol: LOC101928907 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135690.1 | 2 | 523 | |||
| GenBank mRNA | BC028842.1 | - | 2 | 495 | ||
| BC034325.1 | - | 1 | 631 | |||
| DA068710.1 | - | 2 | 523 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000560813.2 | - | 2 | 523 | Chr. 15 - 62060503 - 62061274 [+] on Build GRCh38 | |

LOC101928907
Genomic Map