siRNA Details
Gene Symbol: LOC101928907 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135690.1 | 2 | 560 | |||
GenBank mRNA | BC028842.1 | - | 2 | 532 | ||
BC034325.1 | - | 1 | 668 | |||
DA068710.1 | - | 2 | 560 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000560813.2 | - | 2 | 560 | Chr. 15 - 62060503 - 62061274 [+] on Build GRCh38 |
