siRNA Details
| Gene Symbol: LOC105378349 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_134314.1 | 2 | 431 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000558852.2 | - | 2 | 351 | Chr. 10 - 70939133 - 70944142 [+] on Build GRCh38 | |

LOC105378349
Genomic Map