siRNA Details
| Gene Symbol: LINGO1-AS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120361.1 | 3 | 467 | |||
| NR_120362.1 | 3 | 449 | ||||
| GenBank mRNA | BC038758.1 | - | 3 | 465 | ||
| BI463387.1 | - | 3 | 470 | |||
| HY007517.1 | - | 3 | 438 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000557799.1 | - | 3 | 466 | Chr. 15 - 77660052 - 77668074 [+] on Build GRCh38 | |

LINGO1-AS2
Genomic Map