siRNA Details
| Gene Symbol: LIPC-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120338.1 | 3 | 617 | |||
| GenBank mRNA | AK091917.1 | - | 3 | 617 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000561083.1 | - | 3 | 617 | Chr. 15 - 58434890 - 58498735 [-] on Build GRCh38 | |

LIPC-AS1
Genomic Map