siRNA Details
| Gene Symbol: TMC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120365.1 | 3 | 553 | |||
| GenBank mRNA | BI821075.1 | - | 3 | 468 | ||
| CB134362.1 | - | 3 | 470 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMC3-AS1| Gene Symbol: TMC3-AS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120365.1 | 3 | 553 | |||
| GenBank mRNA | BI821075.1 | - | 3 | 468 | ||
| CB134362.1 | - | 3 | 470 | |||