siRNA Details
Gene Symbol: LOC101929457 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NR_135692.1 | 2 | 2771 | |||
GenBank mRNA | BC062542.1 | - | 2 | 2771 |
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
---|---|---|---|---|---|---|
GENCODE | ENST00000561283.2 | - | 2 | 2826 | Chr. 15 - 77525540 - 77534110 [+] on Build GRCh38 |
