siRNA Details
| Gene Symbol: LOC101929457 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_135692.1 | 2 | 2380 | |||
| GenBank mRNA | BC062542.1 | - | 2 | 2380 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000561283.2 | - | 2 | 2435 | Chr. 15 - 77525540 - 77534110 [+] on Build GRCh38 | |

LOC101929457
Genomic Map