siRNA Details
| Gene Symbol: LOC101929439 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120360.1 | 3 | 1888 | |||
| GenBank mRNA | AK127738.1 | - | 3 | 1888 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000558424.1 | - | 3 | 1888 | Chr. 15 - 76174891 - 76181486 [-] on Build GRCh38 | |

LOC101929439
Genomic Map