siRNA Details
| Gene Symbol: LOC101928134 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NR_120326.1 | 3 | 297 | |||
| NR_120327.1 | 3 | 283 | ||||
| GenBank mRNA | BX095733.1 | - | 3 | 297 | ||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000561458.5 | - | 3 | 298 | Chr. 15 - 33303657 - 33310246 [-] on Build GRCh38 | |

LOC101928134
Genomic Map