siRNA Details
| Gene Symbol: CEP152 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_011521374.2 | XP_011519676.1 | 28 | 5197 | ||
| XM_011521378.2 | XP_011519680.1 | 27 | 5038 | |||
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | Transcript Location | |
|---|---|---|---|---|---|---|
| GENCODE | ENST00000559165.1 | - | 2 | 463 | Chr. 15 - 48729080 - 48729844 [-] on Build GRCh38 | |

CEP152
Genomic Map